EMAIL THIS PAGE TO A FRIEND
W237906 Sigma-Aldrich

Dihydrocarveol

≥96%

  • CAS Number 619-01-2

  • Empirical Formula (Hill Notation) C10H18O

  • Molecular Weight 154.25

  •  FEMA Number 2379

  •  EC Number 210-575-1

  •  Council of Europe no. 2025

  •  MDL number MFCD00001503

  •  PubChem Substance ID 24901034

  •  Flavis number 2.061

Purchase

Properties

grade   Kosher
vapor density   5.3 (vs air)
vapor pressure   0.1 mmHg ( 20 °C)
assay   ≥96%
refractive index   n20/D 1.477 (lit.)
  n20/D 1.479
bp   224-225 °C(lit.)
density   0.924 g/mL at 25 °C
  0.926 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

WGK Germany 
2
RTECS 
OT0175150
Flash Point(F) 
197 °F
Flash Point(C) 
92 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?