• USA Home
  • W239208 - α,α-Dimethylphenethyl acetate

EMAIL THIS PAGE TO A FRIEND
W239208 Sigma-Aldrich

α,α-Dimethylphenethyl acetate

≥98%, FCC, FG

  • CAS Number 151-05-3

  • Empirical Formula (Hill Notation) C12H16O2

  • Molecular Weight 192.25

  •  FEMA Number 2392

  •  EC Number 205-781-3

  •  Council of Europe no. 2077

  •  MDL number MFCD00026196

  •  PubChem Substance ID 24901042

  •  Flavis number 9.227

  •  NACRES NA.21

Purchase

Properties

Related Categories A - D, A-D, Alphabetical Listings, C-D, Certified Food Grade Products,
biological source   synthetic
grade   FG
  Halal
  Kosher
reg. compliance   FCC
  FDA 21 CFR (117)
  FDA 21 CFR (172.515)
  EU Regulation 1334/2008 & 178/2002
assay   ≥98%
refractive index   n20/D 1.4925 (lit.)
bp   250 °C (lit.)
density   0.998 g/mL at 25 °C (lit.)
Documentation   see Safety & Documentation for available documents
Organoleptic   floral; fruity; pear
food allergen   no known allergens
SMILES string   CC(=O)OC(C)(C)Cc1ccccc1
InChI   1S/C12H16O2/c1-10(13)14-12(2,3)9-11-7-5-4-6-8-11/h4-8H,9H2,1-3H3
InChI key   FLUWAIIVLCVEKF-UHFFFAOYSA-N

Description

Packaging

1 kg in poly bottle

5, 10 kg in poly drum

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
SG8100000
Flash Point(F) 
206.6 °F
Flash Point(C) 
97 °C
Protocols & Articles

Articles

Understanding the Complexities of Kosher Ingredients

Introduction What is Kosher Product Considerations for Kosher Certification Kosher's Added Benefit to the Food Industry Sigma-Aldrich's Kosher Experience and Offering
Dr. Luke Grocholl1, Rabbi Gershon Segal

1Dr. Luke Grocholl, Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food & Beverage, Safety industry

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?