• USA Home
  • W239301 - 2-Methyl-1-phenyl-2-propanol

EMAIL THIS PAGE TO A FRIEND
W239301 Sigma-Aldrich

2-Methyl-1-phenyl-2-propanol

98%

Synonym: 1,1-Dimethyl-2-phenylethyl alcohol, Benzyl dimethyl carbinol

  • CAS Number 100-86-7

  • Linear Formula C6H5CH2C(CH3)2OH

  • Molecular Weight 150.22

  •  FEMA Number 2393

  •  Beilstein/REAXYS Number 1855608

  •  EC Number 202-896-0

  •  MDL number MFCD00004465

  •  PubChem Substance ID 329830480

  •  Flavis number 2.035

Purchase

Properties

Related Categories Alphabetical Listings, Flavors and Fragrances, M-N More...
biological source   synthetic
reg. compliance   FDA 21 CFR (172.515)
InChI Key   RIWRBSMFKVOJMN-UHFFFAOYSA-N
assay   98%
refractive index   n20/D 1.514 (lit.)
bp   94-96 °C/10 mmHg(lit.)
mp   23-25 °C (lit.)
density   0.974 g/mL at 25 °C (lit.)
Documentation   see Safety & Documentation for available documents
food allergen   no known allergens
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
SG8050000
Flash Point(F) 
177.8 °F
Flash Point(C) 
81 °C

New Product Offerings
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?