EMAIL THIS PAGE TO A FRIEND
W266604 Sigma-Aldrich

D-Neomenthol

≥99%

Synonym: (+)-Neomenthol, (1S,2S,5R)-(+)-Neomenthol, (1S,2S,5R)-2-Isopropyl-5-methylcyclohexanol, D-Neomenthol, [1S-(1β,2β,5α)]-5-Methyl-2-(1-methylethyl)cyclohexanol

  • CAS Number 2216-52-6

  • Empirical Formula (Hill Notation) C10H20O

  • Molecular Weight 156.27

  •  FEMA Number 2666

  •  Beilstein/REAXYS Number 2037488

  •  EC Number 218-691-4

  •  Council of Europe no. 2028

  •  MDL number MFCD00062980

  •  PubChem Substance ID 24901230

  •  Flavis number 2.063

Purchase

Properties

grade   Kosher
vapor pressure   0.8 mmHg ( 20 °C)
assay   ≥99%
optical activity   [α]24/D +16°, neat
refractive index   n20/D 1.461 (lit.)
bp   95 °C/12 mmHg (lit.)
mp   −22 °C (lit.)
density   0.899 g/mL at 25 °C (lit.)
SMILES string   CC(C)[C@@H]1CC[C@@H](C)C[C@@H]1O
InChI   1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3/t8-,9+,10+/m1/s1
InChI key   NOOLISFMXDJSKH-UTLUCORTSA-N

Description

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
3
RTECS 
OT0450000
Flash Point(F) 
181.4 °F
Flash Point(C) 
83 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?