EMAIL THIS PAGE TO A FRIEND
W279307 Sigma-Aldrich

Nutmeg oil

Synonym: Myristicaceae, Nutmeg oil, East Indian

Purchase

Properties

Related Categories Alphabetical Listings, Essential Oils, Flavors and Fragrances, I-N, Kosher Certified Products,
Quality Level   300
biological source   Myristica fragrans Houtt.
grade   Kosher
reg. compliance   FDA 21 CFR 182.20
optical activity   [α]20/D +10°, neat
origin   Indonesia origin
refractive index   n20/D 1.481 (lit.)
bp   165 °C (lit.)
density   0.89 g/mL at 25 °C (lit.)
Documentation   see Safety & Documentation for available documents
Organoleptic   nutmeg; woody; spicy
food allergen   no known allergens

Description

Packaging

1 kg in glass bottle

Preparation Note

Extraction method: steam distillation

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Respiratory system
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
WGK 2
RTECS 
RJ3694800
Flash Point(F) 
109.4 °F - closed cup
Flash Point(C) 
43 °C - closed cup
Protocols & Articles

Articles

Understanding the Complexities of Kosher Ingredients

Introduction What is Kosher Product Considerations for Kosher Certification Kosher's Added Benefit to the Food Industry Sigma-Aldrich's Kosher Experience and Offering
Dr. Luke Grocholl1, Rabbi Gershon Segal

1Dr. Luke Grocholl, Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food & Beverage, Safety industry

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?