EMAIL THIS PAGE TO A FRIEND
W286001 Sigma-Aldrich

Phenethyl benzoate

≥99%, FG

  • CAS Number 94-47-3

  • Linear Formula C6H5CO2CH2CH2C6H5

  • Molecular Weight 226.27

  •  FEMA Number 2860

  •  EC Number 202-336-5

  •  Council of Europe no. 667

  •  MDL number MFCD00020681

  •  PubChem Substance ID 24901351

  •  Flavis number 9.774

Purchase

Properties

Related Categories Alphabetical Listings, Certified Food Grade Products, Flavors and Fragrances, Kosher Certified Products, N-P,
biological source   synthetic
grade   FG
  Kosher
reg. compliance   EU Regulation 1334/2008 & 872/2012
InChI Key   OSORMYZMWHVFOZ-UHFFFAOYSA-N
assay   ≥99%
refractive index   n20/D 1.56 (lit.)
bp   182 °C/12 mmHg(lit.)
density   1.093 g/mL at 25 °C (lit.)
Documentation   see Safety & Documentation for available documents
Organoleptic   honey; floral; rose
food allergen   no known allergens

Description

Packaging

1 kg in poly bottle

5 kg in poly drum

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
DH6288000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

New Product Offerings
Protocols & Articles

Articles

Understanding the Complexities of Kosher Ingredients

Introduction What is Kosher Product Considerations for Kosher Certification Kosher's Added Benefit to the Food Industry Sigma-Aldrich's Kosher Experience and Offering
Dr. Luke Grocholl1, Rabbi Gershon Segal

1Dr. Luke Grocholl, Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food & Beverage, Safety industry

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food
06732 Timestrip® Seafood

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?