• USA Home
  • W286206 - Phenethyl isobutyrate

EMAIL THIS PAGE TO A FRIEND
W286206 Sigma-Aldrich

Phenethyl isobutyrate

≥98%, FCC, FG

Synonym: 2-Phenylethyl 2-methylpropanoate, 2-Phenylethyl isobutyrate

  • CAS Number 103-48-0

  • Linear Formula (CH3)2CHCO2CH2CH2C6H5

  • Molecular Weight 192.25

  •  FEMA Number 2862

  •  EC Number 203-116-1

  •  Council of Europe no. 302

  •  MDL number MFCD00026441

  •  PubChem Substance ID 24901353

  •  Flavis number 9.427

  •  NACRES NA.21

Purchase

Properties

Quality Level   300
biological source   synthetic
grade   FG
  Kosher
reg. compliance   EU Regulation 1334/2008 & 872/2012
  FCC
  FDA 21 CFR 172.515
assay   ≥98%
refractive index   n20/D 1.4873 (lit.)
bp   250 °C (lit.)
density   0.988 g/mL at 25 °C (lit.)
Documentation   see Safety & Documentation for available documents
Organoleptic   fruity; floral; rose
food allergen   no known allergens
SMILES string   CC(C)C(=O)OCCc1ccccc1
InChI   1S/C12H16O2/c1-10(2)12(13)14-9-8-11-6-4-3-5-7-11/h3-7,10H,8-9H2,1-2H3
InChI key   JDQVBGQWADMTAM-UHFFFAOYSA-N

Description

General description

Phenethyl isobutyrate can be used as a flavoring agent and fragrance ingredient. It is one of the important volatile metabolite formed in the Kluyveromyces lactis broth during fermentation.

Packaging

1 kg in poly bottle

10 kg in poly drum

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 2
RTECS 
NQ5435000
Flash Point(F) 
226.4 °F - closed cup
Flash Point(C) 
108.00 °C - closed cup
Protocols & Articles

Articles

Understanding the Complexities of Kosher Ingredients

Introduction What is Kosher Product Considerations for Kosher Certification Kosher's Added Benefit to the Food Industry Sigma-Aldrich's Kosher Experience and Offering
Dr. Luke Grocholl1, Rabbi Gershon Segal

1Dr. Luke Grocholl, Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food & Beverage, Safety industry

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?