• USA Home
  • W289108 - 2-Phenylpropyl butyrate

EMAIL THIS PAGE TO A FRIEND
W289108 Sigma-Aldrich

2-Phenylpropyl butyrate

≥98%

Synonym: Hydratropyl butyrate

  • CAS Number 80866-83-7

  • Linear Formula CH3CH2CH2CO2CH2CH(C6H5)CH3

  • Molecular Weight 206.28

  •  FEMA Number 2891

  •  EC Number 279-587-2

  •  Council of Europe no. 285

  •  MDL number MFCD00009393

  •  Flavis number 9.057

Purchase

Properties

biological source   synthetic
grade   Kosher
assay   ≥98%
refractive index   n20/D 1.4887 (lit.)
density   0.991 g/mL at 25 °C (lit.)
Organoleptic   apricot; sweet
SMILES string   CCCC(=O)OCC(C)c1ccccc1
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles

Articles

Understanding the Complexities of Kosher Ingredients

Introduction What is Kosher Product Considerations for Kosher Certification Kosher's Added Benefit to the Food Industry Sigma-Aldrich's Kosher Experience and Offering
Dr. Luke Grocholl1, Rabbi Gershon Segal

1Dr. Luke Grocholl, Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food & Beverage, Safety industry

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?