EMAIL THIS PAGE TO A FRIEND
W331401 Sigma-Aldrich

2-Naphthalenethiol

99%

Synonym: 2-Naphthyl mercaptan, Thio-2-naphthol (β)

  • CAS Number 91-60-1

  • Linear Formula C10H7SH

  • Molecular Weight 160.24

  •  FEMA Number 3314

  •  Beilstein/REAXYS Number 636389

  •  EC Number 202-082-5

  •  MDL number MFCD00004086

  •  PubChem Substance ID 329830596

  •  Flavis number 12.033

Purchase

Properties

Related Categories Alphabetical Listings, Flavors and Fragrances, M-N More...
biological source   synthetic
InChI Key   RFCQDOVPMUSZMN-UHFFFAOYSA-N
assay   99%
bp   286 °C(lit.)
mp   79-81 °C (lit.)
Documentation   see Safety & Documentation for available documents
Organoleptic   sulfurous
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN 3335
WGK Germany 
3
RTECS 
QK3930000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

New Product Offerings
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?