• USA Home
  • W339806 - Glyceryl tribenzoate

EMAIL THIS PAGE TO A FRIEND
W339806 Aldrich

Glyceryl tribenzoate

95%

Synonym: Tribenzoin

  • CAS Number 614-33-5

  • Linear Formula C6H5CO2CH(CH2O2CC6H5)2

  • Molecular Weight 404.41

  •  FEMA Number 3398

  •  EC Number 210-379-6

  •  MDL number MFCD00020677

  •  PubChem Substance ID 329830612

  •  Flavis number 9.812

Purchase

Properties

Related Categories Alphabetical Listings, Flavors and Fragrances, G-H More...
biological source   synthetic
InChI Key   HIZCTWCPHWUPFU-UHFFFAOYSA-N
assay   95%
mp   68-72 °C (lit.)
Documentation   see Safety & Documentation for available documents
Organoleptic   green
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?