• USA Home
  • W340000 - Coconut ketone solution

EMAIL THIS PAGE TO A FRIEND
W340000 Sigma-Aldrich

Coconut ketone solution

10 wt. % in ethanol, natural (US)

Synonym: 3-Hepten-2-one

  • CAS Number 1119-44-4

  • Empirical Formula (Hill Notation) C7H12O

  • Molecular Weight 112.17

  •  FEMA Number 3400

  •  Council of Europe no. 11094

  •  MDL number MFCD00015564

  •  PubChem Substance ID 329830613

  •  Flavis number 7.105

  •  NACRES NA.21

Purchase

Properties

Related Categories A-D, Alphabetical Listings, C-D, Certified Natural Products, Flavors and Fragrances,
biological source   botanical
grade   Kosher
  natural (US)
reg. compliance   FDA 21 CFR (117)
InChI Key   JHHZQADGLDKIPM-AATRIKPKSA-N
concentration   10 wt. % in ethanol
refractive index   n20/D 1.370 (lit.)
density   0.805 g/mL at 25 °C (lit.)
Documentation   see Safety & Documentation for available documents
Organoleptic   cheese; coconut; creamy; grassy; green; nutty; earthy; waxy
food allergen   no known allergens

Description

General description

Natural occurence (neat): Roasted hazelnut, green pepper, and hops oil

Application

Useful in most dairy flavors to add greater depth. This ketone will add lingering rich after-notes, to many processed dairy products such as custard, melted butter, creams and cheeses (Brie, Gorgonzola and blue cheeses). Also beneficial in fatty nut applications (brazil and pine nuts, cashews)

Packaging

1 kg in poly bottle

100, 500 g in poly bottle

Biochem/physiol Actions

Odor at 2-5ppm

Taste at 10%

Safety & Documentation

Safety Information

RIDADR 
UN1170 - class 3 - PG 2 - Ethanol, solution
WGK Germany 
3
Flash Point(F) 
57.2 °F
Flash Point(C) 
14 °C

New Product Offerings
Protocols & Articles

Articles

The Complex Regulatory Landscape for Natural Flavor Ingredients

Introduction The Regulatory Landscape US and EU Definitions of Natural Regional Regulatory Variations The Example of Vanillin The Case for Standardization
Mr. Michael Sabisch1, Mr. David Smith2

1Senior Quality Assurance Specialist
2International Sales Coordinator, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food Safety, Oxidations

Understanding the Complexities of Kosher Ingredients

Introduction What is Kosher Product Considerations for Kosher Certification Kosher's Added Benefit to the Food Industry Sigma-Aldrich's Kosher Experience and Offering
Dr. Luke Grocholl1, Rabbi Gershon Segal

1Dr. Luke Grocholl, Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food & Beverage, Safety industry

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food
06732 Timestrip® Seafood

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?