• USA Home
  • W347302 - 2,2,6-Trimethylcyclohexanone

EMAIL THIS PAGE TO A FRIEND
W347302 Sigma-Aldrich

2,2,6-Trimethylcyclohexanone

≥98%

  • CAS Number 2408-37-9

  • Linear Formula (CH3)3C6H7(=O)

  • Molecular Weight 140.22

  •  FEMA Number 3473

  •  Beilstein/REAXYS Number 1905593

  •  EC Number 219-309-9

  •  Council of Europe no. 686

  •  MDL number MFCD00045550

  •  PubChem Substance ID 24901722

  •  Flavis number 7.045

Purchase

Properties

assay   ≥98%
refractive index   n20/D 1.447 (lit.)
bp   178-179 °C (lit.)
density   0.904 g/mL at 25 °C (lit.)
SMILES string   CC1CCCC(C)(C)C1=O
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1224C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
123.8 °F
Flash Point(C) 
51 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?