• USA Home
  • W350516 - 3-Methylbutyl 2-methylbutanoate

EMAIL THIS PAGE TO A FRIEND
W350516 Sigma-Aldrich

3-Methylbutyl 2-methylbutanoate

natural, 70% 3-methylbutyl 2-methylbutanoate basis

Synonym: Isoamyl 2-methylbutyrate

  • CAS Number 27625-35-0

  • Empirical Formula (Hill Notation) C10H20O2

  • Molecular Weight 172.26

  •  FEMA Number 3505

  •  EC Number 248-581-1

  •  Council of Europe no. 10721

  •  MDL number MFCD00048645

  •  PubChem Substance ID 24901745

  •  Flavis number 9.530

Purchase

Properties

Related Categories Alphabetical Listings, Certified Natural Products, Flavors and Fragrances, Halal Certified Products, I - N,
grade   Halal
  Kosher
  natural
assay   70% 3-methylbutyl 2-methylbutanoate basis
impurities   30% 3-methylbutyl 3-methylbutanoate
refractive index   n20/D 1.413 (lit.)
bp   41 °C/1.5 mmHg (lit.)
density   0.857 g/mL at 25 °C (lit.)
Documentation   see Safety & Documentation for available documents
Organoleptic   apple; sweet
food allergen   no known allergens
SMILES string   CCC(C)C(=O)OCCC(C)C
InChI   1S/C10H20O2/c1-5-9(4)10(11)12-7-6-8(2)3/h8-9H,5-7H2,1-4H3
InChI key   VGIRHYHLQKDEPP-UHFFFAOYSA-N

Description

Packaging

1 kg in glass bottle

100 g in glass bottle

4 kg in poly drum

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
WGK 3
Protocols & Articles

Articles

The Complex Regulatory Landscape for Natural Flavor Ingredients

Introduction The Regulatory Landscape US and EU Definitions of Natural Regional Regulatory Variations The Example of Vanillin The Case for Standardization
Mr. Michael Sabisch1, Mr. David Smith2

1Senior Quality Assurance Specialist
2International Sales Coordinator, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food Safety, Oxidations

Understanding the Complexities of Kosher Ingredients

Introduction What is Kosher Product Considerations for Kosher Certification Kosher's Added Benefit to the Food Industry Sigma-Aldrich's Kosher Experience and Offering
Dr. Luke Grocholl1, Rabbi Gershon Segal

1Dr. Luke Grocholl, Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food & Beverage, Safety industry

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?