• USA Home
  • W362107 - 4,5-Dimethyl-2-isobutyl-3-thiazoline

EMAIL THIS PAGE TO A FRIEND
W362107 Sigma-Aldrich

4,5-Dimethyl-2-isobutyl-3-thiazoline

97%

  • CAS Number 65894-83-9

  • Empirical Formula (Hill Notation) C9H17NS

  • Molecular Weight 171.30

  •  FEMA Number 3621

  •  EC Number 265-969-6

  •  MDL number MFCD01680822

  •  PubChem Substance ID 24901814

  •  Flavis number 15.032

Purchase

Properties

biological source   synthetic
assay   97%
refractive index   n20/D 1.4870 (lit.)
bp   208 °C/760 mmHg (lit.)
density   1.487 g/mL at 25 °C (lit.)
Organoleptic   green
SMILES string   CC(C)CC1SC(C)C(C)=N1
InChI   1S/C9H17NS/c1-6(2)5-9-10-7(3)8(4)11-9/h6,8-9H,5H2,1-4H3
InChI key   FDOISHJOXPONIV-UHFFFAOYSA-N
Safety & Documentation

Safety Information

WGK Germany 
2
RTECS 
XJ6642800
Flash Point(F) 
188.6 °F
Flash Point(C) 
87 °C
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?