EMAIL THIS PAGE TO A FRIEND
W369501 Sigma-Aldrich

4-Ethoxyphenol

99%

Synonym: Hydroquinone monoethyl ether

  • CAS Number 622-62-8

  • Linear Formula C2H5OC6H4OH

  • Molecular Weight 138.16

  •  FEMA Number 3695

  •  EC Number 210-748-1

  •  MDL number MFCD00002334

  •  PubChem Substance ID 329830646

  •  Flavis number 4.037

Purchase

Properties

biological source   synthetic
assay   99%
bp   121 °C/9 mmHg (lit.)
mp   64-67 °C (lit.)
Organoleptic   hawthorne
SMILES string   CCOc1ccc(O)cc1
InChI   1S/C8H10O2/c1-2-10-8-5-3-7(9)4-6-8/h3-6,9H,2H2,1H3
InChI key   LKVFCSWBKOVHAH-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
SL3790000
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?