• USA Home
  • W382809 - 2,4-Dimethylanisole

EMAIL THIS PAGE TO A FRIEND
W382809 Sigma-Aldrich

2,4-Dimethylanisole

≥97%

  • CAS Number 6738-23-4

  • Linear Formula (CH3)2C6H3OCH3

  • Molecular Weight 136.19

  •  FEMA Number 3828

  •  EC Number 229-794-9

  •  MDL number MFCD00008377

  •  PubChem Substance ID 24901900

  •  Flavis number 4.063

Purchase

Properties

biological source   synthetic
InChI Key   UJCFZCTTZWHRNL-UHFFFAOYSA-N
assay   ≥97%
refractive index   n20/D 1.514 (lit.)
bp   191 °C (lit.)
density   0.973 g/mL at 25 °C (lit.)
Organoleptic   spicy

Description

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
145.4 °F
Flash Point(C) 
63 °C
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?