• USA Home
  • W392200 - trans,trans-2,4-Hexadien-1-ol

EMAIL THIS PAGE TO A FRIEND
W392200 Sigma-Aldrich

trans,trans-2,4-Hexadien-1-ol

≥97%, stabilized, FG

Synonym: (E,E)-2,4-Hexadien-1-ol, Sorbic alcohol

  • CAS Number 17102-64-6

  • Linear Formula CH3CH=CHCH=CHCH2OH

  • Molecular Weight 98.14

  •  FEMA Number 3922

  •  Beilstein/REAXYS Number 1719722

  •  MDL number MFCD00002925

  •  PubChem Substance ID 24901938

  •  Flavis number 2.162

Purchase

Properties

Related Categories Alphabetical Listings, Certified Food Grade Products, Flavors and Fragrances, G-H, H-J,
biological source   synthetic
grade   FG
  Kosher
reg. compliance   FDA 21 CFR (110)
  EU Regulation 1334/2008 & 872/2012
vapor density   >1 (vs air)
InChI Key   MEIRRNXMZYDVDW-MQQKCMAXSA-N
assay   ≥97%
contains   α- tocopherol (synthetic as stabilizer)
refractive index   n20/D 1.5 (lit.)
bp   80 °C/12 mmHg(lit.)
mp   28-33 °C (lit.)
density   0.871 g/mL at 25 °C (lit.)
Documentation   see Safety & Documentation for available documents
Organoleptic   fresh; green; herbaceous
food allergen   no known allergens

Description

General description

trans,trans-2,4-Hexadien-1-ol is an α,β-unsaturated aldehyde that can be used as a flavor ingredient.

Packaging

100 g in glass bottle

25 g in poly bottle

Biochem/physiol Actions

Odor at 1%

Taste at 10-40 ppm

Other Notes

Natural occurrence: Raw and cooked asparagus, naranjilla fruit.

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
2

New Product Offerings
Protocols & Articles

Articles

Understanding the Complexities of Kosher Ingredients

Introduction What is Kosher Product Considerations for Kosher Certification Kosher's Added Benefit to the Food Industry Sigma-Aldrich's Kosher Experience and Offering
Dr. Luke Grocholl1, Rabbi Gershon Segal

1Dr. Luke Grocholl, Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food & Beverage, Safety industry

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food
06732 Timestrip® Seafood

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?