• USA Home
  • W396508 - DL-1-Amino-2-propanol

EMAIL THIS PAGE TO A FRIEND
W396508 Sigma-Aldrich

DL-1-Amino-2-propanol

≥90%

Synonym: (±)-Isopropanolamine, (±)-1-Amino-2-propanol

  • CAS Number 78-96-6

  • Linear Formula CH3CH(OH)CH2NH2

  • Molecular Weight 75.11

  •  FEMA Number 3965

  •  Beilstein/REAXYS Number 605275

  •  EC Number 201-162-7

  •  MDL number MFCD00008139

  •  PubChem Substance ID 329830669

  •  NACRES NA.21

Purchase

Properties

Related Categories A-B, Alphabetical Listings, Flavors and Fragrances More...
Quality Level   300
biological source   synthetic
reg. compliance   FDA 21 CFR 117
vapor density   2.6 (vs air)
vapor pressure   <1 mmHg ( 20 °C)
assay   ≥90%
impurities   <10% 2-amino-1-propanol
refractive index   n20/D 1.4478 (lit.)
bp   160 °C (lit.)
mp   −2 °C (lit.)
density   0.973 g/mL at 25 °C (lit.)
Documentation   see Safety & Documentation for available documents
Organoleptic   fishy
food allergen   no known allergens
SMILES string   CC(O)CN
InChI   1S/C3H9NO/c1-3(5)2-4/h3,5H,2,4H2,1H3
InChI key   HXKKHQJGJAFBHI-UHFFFAOYSA-N

Description

Packaging

1 kg in poly bottle

10, 25 kg in poly drum

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2735PSN1 8 / PGII
WGK Germany 
WGK 1
RTECS 
UA5775000
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?