EMAIL THIS PAGE TO A FRIEND
W402206 Sigma-Aldrich

6-Undecanone

Synonym: Diamyl ketone, Dipentyl ketone

  • CAS Number 927-49-1

  • Linear Formula CH3(CH2)4CO(CH2)4CH3

  • Molecular Weight 170.29

  •  FEMA Number 4022

  •  EC Number 213-150-9

  •  MDL number MFCD00009516

  •  PubChem Substance ID 24901973

  •  Flavis number 7.249

Purchase

Properties

Related Categories Alphabetical Listings, Flavors and Fragrances, Q-Z More...
biological source   synthetic
refractive index   n20/D 1.4270 (lit.)
bp   228 °C (lit.)
mp   14.6 °C (lit.)
density   0.831 g/mL at 25 °C (lit.)
Organoleptic   odorless
SMILES string   CCCCCC(=O)CCCCC
InChI   1S/C11H22O/c1-3-5-7-9-11(12)10-8-6-4-2/h3-10H2,1-2H3
InChI key   ZPQAKYPOZRXKFA-UHFFFAOYSA-N

Description

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
YQ2828000
Flash Point(F) 
190.4 °F
Flash Point(C) 
88 °C
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?