EMAIL THIS PAGE TO A FRIEND
W406503 Sigma-Aldrich

2-Propylpyridine

≥97%

Synonym: Conyrine

  • CAS Number 622-39-9

  • Empirical Formula (Hill Notation) C8H11N

  • Molecular Weight 121.18

  •  FEMA Number 4065

  •  EC Number 210-732-4

  •  MDL number MFCD00006368

  •  PubChem Substance ID 329830673

  •  Flavis number 14.164

Purchase

Properties

biological source   synthetic
assay   ≥97%
refractive index   n20/D 1.493
bp   169-171 °C
density   0.912 g/mL at 25 °C
Documentation   see Safety & Documentation for available documents
Organoleptic   green; minty; floral
food allergen   no known allergens
SMILES string   CCCc1ccccn1
InChI   1S/C8H11N/c1-2-5-8-6-3-4-7-9-8/h3-4,6-7H,2,5H2,1H3
InChI key   OIALIKXMLIAOSN-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
132.8 °F
Flash Point(C) 
56 °C
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?