EMAIL THIS PAGE TO A FRIEND
W425001 Sigma-Aldrich

Piperazine

99%

Synonym: 1,4-Diazacyclohexane, Diethylenediamine

  • CAS Number 110-85-0

  • Empirical Formula (Hill Notation) C4H10N2

  • Molecular Weight 86.14

  •  FEMA Number 4250

  •  Beilstein/REAXYS Number 102555

  •  EC Number 203-808-3

  •  MDL number MFCD00005953

  •  PubChem Substance ID 329830688

  •  Flavis number 14.141

Purchase

Properties

biological source   synthetic
vapor pressure   0.8 mmHg ( 20 °C)
assay   99%
expl. lim.   14 %
bp   145-146 °C (lit.)
mp   109-112 °C (lit.)
solubility   H2O: 0.9 g/L at 20 °C
Organoleptic   camphoraceous
SMILES string   C1CNCCN1
InChI   1S/C4H10N2/c1-2-6-4-3-5-1/h5-6H,1-4H2
InChI key   GLUUGHFHXGJENI-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2579 8 / PGIII
WGK Germany 
1
RTECS 
TK7800000
Flash Point(F) 
228.2 °F
Flash Point(C) 
109 °C
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?