• USA Home
  • W442500 - Isopropyl isothiocyanate

EMAIL THIS PAGE TO A FRIEND
W442500 Aldrich

Isopropyl isothiocyanate

97%

Synonym: Isopropyl mustard oil

Purchase

Properties

biological source   synthetic
InChI Key   VHBFEIBMZHEWSX-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.493(lit.)
bp   29-30 °C/10 mmHg(lit.)
density   0.948 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
104 °F
Flash Point(C) 
40 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?