• USA Home
  • W463102 - 2-Ethyl-3-methylthiopyrazine

EMAIL THIS PAGE TO A FRIEND
W463102 Sigma-Aldrich

2-Ethyl-3-methylthiopyrazine

97%

  • CAS Number 72987-62-3

  • Empirical Formula (Hill Notation) C7H10N2S

  • Molecular Weight 154.23

  •  FEMA Number 4631

  •  EC Number 277-191-4

  •  MDL number MFCD00038027

  •  PubChem Substance ID 329830722

  •  NACRES NA.21

Purchase

Properties

Related Categories Alphabetical Listings, E-F, Flavors and Fragrances More...
Quality Level   300
biological source   synthetic
reg. compliance   FDA 21 CFR 117
assay   97%
Documentation   see Safety & Documentation for available documents
Organoleptic   roasted
food allergen   no known allergens
SMILES string   CCc1nccnc1SC
InChI   1S/C7H10N2S/c1-3-6-7(10-2)9-5-4-8-6/h4-5H,3H2,1-2H3
InChI key   XYHPPOMSLGJAAM-UHFFFAOYSA-N

Description

Packaging

100, 500 g in poly bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?