• USA Home
  • W463305 - 2-Ethoxy-3-ethylpyrazine

EMAIL THIS PAGE TO A FRIEND
W463305 Sigma-Aldrich

2-Ethoxy-3-ethylpyrazine

97%

  • CAS Number 35243-43-7

  • Empirical Formula (Hill Notation) C8H12N2O

  • Molecular Weight 152.19

  •  FEMA Number 4633

  •  MDL number MFCD00038026

  •  PubChem Substance ID 329830723

Purchase

Properties

biological source   synthetic
assay   97%
refractive index   n20/D 1.493
density   1.011 g/mL at 25 °C
Documentation   see Safety & Documentation for available documents
Organoleptic   potato
food allergen   no known allergens
SMILES string   CCOc1nccnc1CC
InChI   1S/C8H12N2O/c1-3-7-8(11-4-2)10-6-5-9-7/h5-6H,3-4H2,1-2H3
InChI key   TYSHGGVMHVIOMH-UHFFFAOYSA-N

Description

Packaging

100, 500 g in poly bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
159.8 °F
Flash Point(C) 
71 °C
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?