• USA Home
  • W503401 - 2,4-Dimethylthiazole

EMAIL THIS PAGE TO A FRIEND
W503401 Sigma-Aldrich

2,4-Dimethylthiazole

≥95%, FG

  • CAS Number 541-58-2

  • Empirical Formula (Hill Notation) C5H7NS

  • Molecular Weight 113.18

  •  EC Number 208-786-9

  •  MDL number MFCD00014509

  •  PubChem Substance ID 24902013

  •  Flavis number 15.062

Purchase

Properties

Related Categories C-D, Certified Food Grade Products, Flavors and Fragrances More...
biological source   synthetic
grade   FG
reg. compliance   EU Regulation 1334/2008 & 872/2012
InChI Key   OBSLLHNATPQFMJ-UHFFFAOYSA-N
assay   ≥95%
refractive index   n20/D 1.509 (lit.)
bp   144-146 °C/719 mmHg(lit.)
density   1.056 g/mL at 25 °C (lit.)
Documentation   see Safety & Documentation for available documents
Organoleptic   coffee; meaty
food allergen   no known allergens

Description

Packaging

100 g in glass bottle

25 g in poly bottle

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
RTECS 
XJ4375000
Flash Point(F) 
109.4 °F
Flash Point(C) 
43 °C

New Product Offerings
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food
06732 Timestrip® Seafood

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?