EMAIL THIS PAGE TO A FRIEND
W506400 Sigma-Aldrich

o-Anisaldehyde

≥97%

Synonym: 2-Methoxybenzaldehyde, o-Anisaldehyde, Salicylaldehyde methyl ether

  • CAS Number 135-02-4

  • Linear Formula CH3OC6H4CHO

  • Molecular Weight 136.15

  •  FEMA Number 4077

  •  Beilstein/REAXYS Number 606301

  •  EC Number 205-171-7

  •  MDL number MFCD00003308

  •  PubChem Substance ID 329830740

  •  Flavis number 5.129

Purchase

Properties

biological source   synthetic
grade   Kosher
assay   ≥97%
bp   238 °C (lit.)
mp   34-40 °C (lit.)
density   1.127 g/mL at 25 °C (lit.)
Organoleptic   floral; sweet
SMILES string   [H]C(=O)c1ccccc1OC
InChI   1S/C8H8O2/c1-10-8-5-3-2-4-7(8)6-9/h2-6H,1H3
InChI key   PKZJLOCLABXVMC-UHFFFAOYSA-N

Description

Biochem/physiol Actions

Naturally-occurring aromatic aldehyde with acaricidal activity. May condense with L-tryptophan in foods to form the corresponding phenolic tetrahydro-β-carboline-3-carboxylic acid which is a potent antioxidant.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
BZ2610000
Flash Point(F) 
244.4 °F
Flash Point(C) 
118 °C
Protocols & Articles

Articles

Understanding the Complexities of Kosher Ingredients

Introduction What is Kosher Product Considerations for Kosher Certification Kosher's Added Benefit to the Food Industry Sigma-Aldrich's Kosher Experience and Offering
Dr. Luke Grocholl1, Rabbi Gershon Segal

1Dr. Luke Grocholl, Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food & Beverage, Safety industry

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?