• USA Home
  • W506702 - Methyl phenoxyacetate

EMAIL THIS PAGE TO A FRIEND
W506702 Sigma-Aldrich

Methyl phenoxyacetate

≥99%

Purchase

Properties

biological source   synthetic
assay   ≥99%
refractive index   n20/D 1.514 (lit.)
bp   243 °C (lit.)
density   1.149 g/mL at 25 °C (lit.)
SMILES string   COC(=O)COc1ccccc1
InChI   1S/C9H10O3/c1-11-9(10)7-12-8-5-3-2-4-6-8/h2-6H,7H2,1H3
InChI key   BZCKRPHEZOHHBK-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?