• USA Home
  • W510548 - Benzyl isothiocyanate

EMAIL THIS PAGE TO A FRIEND
W510548 Sigma-Aldrich

Benzyl isothiocyanate

98%

Synonym: Benzyl mustard oil, Isothiocyanotaomethylbenzene

  • CAS Number 622-78-6

  • Linear Formula C6H5CH2NCS

  • Molecular Weight 149.21

  •  Beilstein/REAXYS Number 386135

  •  EC Number 210-753-9

  •  MDL number MFCD00004819

  •  PubChem Substance ID 24902055

  •  Flavis number 12.102

Purchase

Properties

InChI Key   MDKCFLQDBWCQCV-UHFFFAOYSA-N
assay   98%
color   yellow
refractive index   n20/D 1.601 (lit.)
bp   242-243 °C(lit.)
density   1.125 g/mL at 25 °C (lit.)
bulk density   1.125 g/mL
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 3334
WGK Germany 
3
RTECS 
NX8250000
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

W442800 Benzyl isothiocyanate, 98%, FG

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?