• USA Home
  • W510718 - 5α-Androst-16-en-3α-ol

EMAIL THIS PAGE TO A FRIEND
W510718 Sigma-Aldrich

5α-Androst-16-en-3α-ol

≥98%

Synonym: 16-(5α)Androsten-3α-ol, 3α-Hydroxy-5α-androst-16-ene

Purchase

Properties

biological source   synthetic
assay   ≥98%
optical activity   [α]20/D +19°, c = 1 in ethanol
drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
mp   140-141 °C (lit.)
Documentation   see Safety & Documentation for available documents
Organoleptic   faint
food allergen   no known allergens
SMILES string   C[C@]12CC[C@H]3[C@@H](CC[C@H]4C[C@H](O)CC[C@]34C)[C@@H]1CC=C2
InChI   1S/C19H30O/c1-18-9-3-4-16(18)15-6-5-13-12-14(20)7-11-19(13,2)17(15)8-10-18/h3,9,13-17,20H,4-8,10-12H2,1-2H3/t13-,14+,15-,16-,17-,18-,19-/m0/s1
InChI key   KRVXMNNRSSQZJP-PHFHYRSDSA-N

Description

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?