EMAIL THIS PAGE TO A FRIEND
W512702 Sigma-Aldrich

Tridecanal

≥95%

Synonym: Tridecyl aldehyde

Purchase

Properties

Related Categories Alphabetical Listings, Flavors and Fragrances, Halal Certified Products, O - Z, Q-Z More...
biological source   synthetic
grade   Halal
  Kosher
assay   ≥95%
contains   may contain up to 0.50% alpha-tocopherol, synthetic as stabilzer
refractive index   n20/D 1.438 (lit.)
bp   132-136 °C/8 mmHg (lit.)
density   0.835 g/mL at 25 °C (lit.)
Documentation   see Safety & Documentation for available documents
food allergen   no known allergens
SMILES string   CCCCCCCCCCCCC=O
InChI   1S/C13H26O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14/h13H,2-12H2,1H3
InChI key   BGEHHAVMRVXCGR-UHFFFAOYSA-N

Description

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles

Articles

Understanding the Complexities of Kosher Ingredients

Introduction What is Kosher Product Considerations for Kosher Certification Kosher's Added Benefit to the Food Industry Sigma-Aldrich's Kosher Experience and Offering
Dr. Luke Grocholl1, Rabbi Gershon Segal

1Dr. Luke Grocholl, Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food & Beverage, Safety industry

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

W433500 Tridecanal, ≥95%, stabilized

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?