• USA Home
  • W514306 - cis-3-Hexenyl crotonate

EMAIL THIS PAGE TO A FRIEND
W514306 Sigma-Aldrich

cis-3-Hexenyl crotonate

≥97%

  • CAS Number 65405-80-3

  • Linear Formula CH3CH=CHCO2CH2CH2CH=CHC2H5

  • Molecular Weight 168.23

  •  EC Number 265-746-3

  •  MDL number MFCD00059903

  •  Flavis number 9.566

Purchase

Properties

biological source   synthetic
InChI Key   KITGYVIOYOCIIE-QNMAEOQASA-N
assay   ≥97%
contains   may contain up to 0.50% alpha-tocopherol, synthetic as stabilzer
refractive index   n20/D 1.455 (lit.)
bp   107 °C/25 mmHg(lit.)
density   0.91 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
201.2 °F
Flash Point(C) 
94 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?