EMAIL THIS PAGE TO A FRIEND
W517607 Sigma-Aldrich

Acetoacetanilide

≥98%

  • CAS Number 102-01-2

  • Linear Formula CH3COCH2CONHC6H5

  • Molecular Weight 177.20

  •  Beilstein Registry Number 473419

  •  EC Number 202-996-4

  •  MDL number MFCD00008780

  •  PubChem Substance ID 24901924

Purchase

Properties

Related Categories A-B, Alphabetical Listings, Flavors and Fragrances More...
biological source   synthetic
vapor density   6.1 (vs air)
InChI Key   DYRDKSSFIWVSNM-UHFFFAOYSA-N
assay   ≥98%
autoignition temp.   843 °F
mp   83-88 °C (lit.)
Documentation   see Safety & Documentation for available documents
Organoleptic   faint

Description

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
AK4200000
Flash Point(F) 
323.6 °F
Flash Point(C) 
162 °C
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food
06732 Timestrip® Seafood

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?