EMAIL THIS PAGE TO A FRIEND
W525901 Sigma-Aldrich

Coumarin, Japan

Synonym: 1,2-Benzopyrone, 1-Benzopyran-2-one, 2H-Chromen-2-one, Coumarin

  • CAS Number 91-64-5

  • Empirical Formula (Hill Notation) C9H6O2

  • Molecular Weight 146.14

  •  Beilstein/REAXYS Number 383644

  •  EC Number 202-086-7

  •  MDL number MFCD00006850

Purchase

Properties

biological source   synthetic
vapor pressure   0.01 mmHg ( 47 °C)
origin   Japan origin
bp   298 °C (lit.)
mp   68-73 °C (lit.)
SMILES string   O=C1Oc2ccccc2C=C1
InChI   1S/C9H6O2/c10-9-6-5-7-3-1-2-4-8(7)11-9/h1-6H
InChI key   ZYGHJZDHTFUPRJ-UHFFFAOYSA-N

Description

Features and Benefits

Freshly mown hay.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
GN4200000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

W526509 Coumarin, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?