• USA Home
  • W6446 - Methacetin-(methoxy-13C)

EMAIL THIS PAGE TO A FRIEND
W6446 Sigma-Aldrich

Methacetin-(methoxy-13C)

99 atom % 13C, MBT grade

Synonym: N-(4-Methoxy-13C-phenyl)acetamide

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service.
Purchase

Properties

Related Categories Alphabetical Listings, M, Stable Isotopes, cGMP Products
isotopic purity   99 atom % 13C
mp   128-130 °C (lit.)
mass shift   M+1
SMILES string   CC(=O)Nc1ccc(O[13CH3])cc1
InChI   1S/C9H11NO2/c1-7(11)10-8-3-5-9(12-2)6-4-8/h3-6H,1-2H3,(H,10,11)/i2+1
InChI key   XVAIDCNLVLTVFM-VQEHIDDOSA-N

Description

Packaging

This product is available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?