• USA Home
  • Z540390 - Aldrich® Spectral Viewer
EMAIL THIS PAGE TO A FRIEND
Z540390 Sigma-Aldrich
Aldrich® Spectral Viewer
FT-IR, 2002 library, network version
Purchase
Description
General description
The Aldrich Spectral Viewer is an electronic reference book on CD-ROM that contains thousands of spectra from the Aldrich spectral libraries. This easy-to-use software program is more powerful than a printed book, allowing text and data field searching and the manipulation, printing, and exporting of spectra.
Currently, we offer one FT-IR library (over 11,000 compounds), two FT-NMR libraries (over 15,000 compounds), and one ATR-IR library (over 18,000 compounds) to suit your needs. The libraries can be used individually or can be combined to create a powerful, electronic reference. A single user interface handles IR, NMR, and ATR-IR spectra, simplifying the use of the software.
In addition, the Spectral Viewer is expandable so that when new spectral libraries are released, they can be easily added to the software. A student/instructional version is also available with a limited library of FT-NMR, FT-IR, and ATR-IR spectra. The demonstration version is also available with a complete version of the software and a small demonstration database of FT-NMR, FT-IR, and ATR-IR spectra.
For more information on a particular spectral library, please see the individual product listing.
Other Notes
11,000 compounds
System Requirements
300MB hard disk space (server), 30MB hard disk space (client)
Pentium 200MHz, Windows 95 / 98 / NT4-SP5 / 2000 / ME / XP / Vista / 7, CD-ROM drive, 64MB RAM, disk space will vary with libraries installed.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Spectral Viewer is a trademark of Sigma-Aldrich Co. LLC
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?