• USA Home
  • Z541591 - Aldrich® Spectral Viewer
EMAIL THIS PAGE TO A FRIEND
Z541591 Aldrich
Aldrich® Spectral Viewer
Demonstration program
Purchase
Description
General description
The Aldrich Spectral Viewer is an electronic reference book on CD-ROM that contains thousands of spectra from the Aldrich spectral libraries. This easy-to-use software program is more powerful than a printed book, allowing text and data field searching and the manipulation, printing, and exporting of spectra.
Currently, we offer one FT-IR library (over 11,000 compounds), two FT-NMR libraries (over 15,000 compounds), and one ATR-IR library (over 18,000 compounds) to suit your needs. The libraries can be used individually or can be combined to create a powerful, electronic reference. A single user interface handles IR, NMR, and ATR-IR spectra, simplifying the use of the software.
In addition, the Spectral Viewer is expandable so that when new spectral libraries are released, they can be easily added to the software. A student/instructional version is also available with a limited library of FT-NMR, FT-IR, and ATR-IR spectra. The demonstration version is also available with a complete version of the software and a small demonstration database of FT-NMR, FT-IR, and ATR-IR spectra.
For more information on a particular spectral library, please see the individual product listing.
Other Notes
The Spectral Viewer demonstration version contains a small database of compounds with FT-IR and/or FT-NMR spectra to demonstrate the programs functions and insure the examples in the tutorial operate correctly, but is otherwise identical to the full version.
System Requirements
25MB hard disk space (server), 30MB hard disk space (client)
Pentium 200MHz, Windows 95 / 98 / NT4-SP5 / 2000 / ME / XP / Vista / 7, CD-ROM drive, 64MB RAM, disk space will vary with libraries installed.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Spectral Viewer is a trademark of Sigma-Aldrich Co. LLC
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Frequently Asked Questions
Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.
How do I get lot-specific information or a Certificate of Analysis?
The lot specific COA document can be found by entering the lot number above under the "Documents" section.
How do I find price and availability?
There are several ways to find pricing and availability for our products.Once you log onto our website, you will find the price and availability displayed on the product detail page.You can contact any of our Customer Sales and Service offices to receive a quote. USA customers: 1-800-325-3010 orview local office numbers.
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Are customers able to order a demo version of Product No. Z541591, Spectral Viewer?
Actually, Product No. Z541591 is a Demo Program of the full program Aldrich Spectral Viewer.
Is Product No. Z541591,Spectral Viewer,a high resolution database?
No, product Z541591 is not high resolution machine readable data.This product is an electronic reference works that contains a picture image of the spectra.This electronic reference book is on CD-ROM and contains thousands of actual spectra from the Aldrich spectral libraries.
Does the Spectral Viewer (e.g., Product No. Z541591) allow you to search by CAS number, or Molecular Formula?
Yes, the Spectral Viewer allows you to text search by chemical name, Aldrich Catalog Number, CAS Number, and Molecular Formula.Advanced data field searching on Chemical Category and physical properties such as boiling point, melting point, density, flash point, molecular weight, and refractive index are also available for your convenience.
Can you search by inputting Spectral Images with Product No. Z541591?
No, the Spectral Viewer is not searchable by inputting Spectral images.
Is the Spectral Viewer, Product Z541591,expandable?
Yes, the Spectral Viewer is expandable. As new libraries become available, you are encouraged to add these to your collection. There is a minimal fee and no subscription or maintenance fees are required.
My question is not addressed here, how can I contact Technical Service for assistance?
Ask a Scientist here.
Show more questions
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?