EMAIL THIS PAGE TO A FRIEND
A78403 Sigma-Aldrich

4-Aminopyridine

98%

Synonym: 4-Pyridinamine, 4-Pyridylamine, 4AP, Fampridine

  • CAS Number 504-24-5

  • Empirical Formula (Hill Notation) C5H6N2

  • Molecular Weight 94.11

  •  Beilstein Registry Number 105782

  •  EC Number 207-987-9

  •  MDL number MFCD00006439

  •  PubChem Substance ID 24891285

Purchase

Properties

Related Categories Building Blocks, C5, Cell Biology, Cell Signaling and Neuroscience, Chemical Synthesis,
InChI Key   NUKYPUAOHBNCPY-UHFFFAOYSA-N
assay   98%
bp   273 °C(lit.)
mp   155-158 °C (lit.)
solubility   H2O: 50 mg/mL, clear, colorless
Gene Information   mouse ... Kcna3(16491)
rat ... Grin2a(24409)

Description

Packaging

25, 100 g in poly bottle

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2671 6.1 / PGII
WGK Germany 
3
RTECS 
US1750000

Meet Synthia - Retrosynthesis Sofware

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?