• USA Home
  • ALD00414 - 4-Nitrophenyl fluorosulfate

EMAIL THIS PAGE TO A FRIEND
ALD00414 Sigma-Aldrich

4-Nitrophenyl fluorosulfate

95%

Synonym: 4-Nitrophenyl ester sulfuryl fluoride

  • CAS Number 51451-34-4

  • Empirical Formula (Hill Notation) C6H4FNO5S

  • Molecular Weight 221.16

  •  PubChem Substance ID 329772670

Purchase

Description

Packaging

500 mg in glass bottle

Application

The following aryl fluorosulfate can be utilized as a cross-coupling partner in Suzuki-Miyaura reactions. The standard reaction conditions for these palladium-catalyzed C-C bond formations are ligand-free and can be performed in water, at room temperature and are not air sensitive.

Other Notes

Palladium-Catalyzed, Ligand-Free Suzuki Reaction in Water Using Aryl Fluorosulfates

Safety & Documentation

Safety Information

RIDADR 
UN 3265 8 / PGII

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles

Articles

Barry Sharpless Group – Professor Product Portal

From our library of Articles, Sigma-Aldrich presents Barry Sharpless Group – Professor Product Portal
Keywords: Asymmetric synthesis, Building blocks, Chemical reactions, Click chemistry, Oxidations, Solvents, transformation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?