• USA Home
  • BR43028 - BRAND® glass bottle-top tilt pipette dispenser

EMAIL THIS PAGE TO A FRIEND
BR43028

BRAND® glass bottle-top tilt pipette dispenser

volume 50 mL, with bottle

Synonym: Automatic pipettes

  •  NACRES NB.01

Purchase

Properties

Related Categories BRAND, BRAND Pipettes, Pipettors, and Tips, Labware, Liquid Handling Equipment, Manufacturer Browser,
feature   Calibrated to deliver (TD, Ex)
  with bottle
mfr. no.   BRAND, 43028
volume   50 mL
color   green markings
fitting   (29/32)

Description

General description

Calibrated to deliver (TD, Ex). Inscriptions in green enamel. The automatic dispenser is particularly well-suited for dispensing suspensions. Delivvery range approximately +/- 10%.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?