• USA Home
  • C2409 - Carbamoylcholine chloride

EMAIL THIS PAGE TO A FRIEND
C2409 Sigma-Aldrich

Carbamoylcholine chloride

99%

Synonym: (2-Hydroxyethyl)trimethylammonium chloride carbamate, Carbachol, Carbamylcholine chloride

  • CAS Number 51-83-2

  • Linear Formula NH2COOCH2CH2N(Cl)(CH3)3

  • Molecular Weight 182.65

  •  Beilstein/REAXYS Number 3917459

  •  EC Number 200-127-3

  •  MDL number MFCD00012011

  •  PubChem Substance ID 24892328

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   AIXAANGOTKPUOY-UHFFFAOYSA-N
assay   99%
mp   210 °C (dec.) (lit.)
solubility   H2O: 50 mg/mL, clear, colorless
Gene Information   human ... CHRM1(1128), CHRM2(1129), CHRM3(1131), CHRM4(1132), CHRM5(1133)
mouse ... Chrm1(12669)
rat ... Chrm1(25229), Chrm2(81645), Chrm3(24260), Chrna2(170945), Chrna3(25101), Chrna4(25590)
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
RTECS 
GA0875000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?