• USA Home
  • CBR00650 - 1-(4-Propyl-4H-1,2,4-triazol-3-yl)ethanamine dihydrochloride

EMAIL THIS PAGE TO A FRIEND
CBR00650 Sigma-Aldrich

1-(4-Propyl-4H-1,2,4-triazol-3-yl)ethanamine dihydrochloride

AldrichCPR

  • Empirical Formula (Hill Notation) C7H16Cl2N4

  • Molecular Weight 227.13

  •  MDL number MFCD13186211

  •  PubChem Substance ID 329775928

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Halogenated Heterocycles, Heterocyclic Building Blocks, Triazoles More...
InChI Key   FOUIXVIFXXONEL-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Legal Information

Product of ChemBridge Corp.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?