• USA Home
  • CBR00889 - (3-Methoxy-2,2-dimethylpropyl)amine hydrochloride

EMAIL THIS PAGE TO A FRIEND
CBR00889 Sigma-Aldrich

(3-Methoxy-2,2-dimethylpropyl)amine hydrochloride

AldrichCPR

  • Empirical Formula (Hill Notation) C6H16ClNO

  • Molecular Weight 153.65

  •  MDL number MFCD12031492

  •  PubChem Substance ID 329776161

Purchase

Properties

Related Categories Amine Salts, Building Blocks, C2 to C7, Chemical Synthesis, Ethers,
SMILES string   NCC(C)(C)COC.[H]Cl
InChI   1S/C6H15NO.ClH/c1-6(2,4-7)5-8-3;/h4-5,7H2,1-3H3;1H
InChI key   IXJYFALFWJVGKE-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Legal Information

Product of ChemBridge Corp.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?