• USA Home
  • CBR01249 - 5-Chloro-3-phenyl-1H-indole-2-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
CBR01249 Sigma-Aldrich

5-Chloro-3-phenyl-1H-indole-2-carboxylic acid

AldrichCPR

Purchase

Properties

Related Categories Building Blocks, C13 to C27, Chemical Synthesis, Halogenated Heterocycles, Heterocyclic Building Blocks,
SMILES string   OC(=O)c1[nH]c2ccc(Cl)cc2c1-c3ccccc3
InChI   1S/C15H10ClNO2/c16-10-6-7-12-11(8-10)13(14(17-12)15(18)19)9-4-2-1-3-5-9/h1-8,17H,(H,18,19)
InChI key   WQRNPWUZAVBEBC-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?