• USA Home
  • CBR01526 - 3-Amino-2-piperidinone hydrochloride

EMAIL THIS PAGE TO A FRIEND
CBR01526 Sigma-Aldrich

3-Amino-2-piperidinone hydrochloride

AldrichCPR

  • Empirical Formula (Hill Notation) C5H11ClN2O

  • Molecular Weight 150.61

  •  MDL number MFCD09028970

  •  PubChem Substance ID 329776773

Purchase

Properties

Related Categories Building Blocks, C5 to C7, Chemical Synthesis, Halogenated Heterocycles, Heterocyclic Building Blocks,
SMILES string   Cl.NC1CCCNC1=O
InChI   1S/C5H10N2O.ClH/c6-4-2-1-3-7-5(4)8;/h4H,1-3,6H2,(H,7,8);1H
InChI key   NLAYLURYAOXTTE-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?