• USA Home
  • CDS000480 - 3-Methyl-2-butenyl benzoate

EMAIL THIS PAGE TO A FRIEND
CDS000480 Sigma-Aldrich

3-Methyl-2-butenyl benzoate

AldrichCPR

  • Empirical Formula (Hill Notation) C12H14O2

  • Molecular Weight 190.24

  •  MDL number MFCD00036570

  •  PubChem Substance ID 329777816

Purchase

Properties

Related Categories Building Blocks, C12 to C63, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   INVWRXWYYVMFQC-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis


Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?