• USA Home
  • CDS000826 - (1R,2R)-(-)-2-Benzyloxycyclohexyl isocyanate

EMAIL THIS PAGE TO A FRIEND
CDS000826 Sigma-Aldrich

(1R,2R)-(-)-2-Benzyloxycyclohexyl isocyanate

AldrichCPR

  • Empirical Formula (Hill Notation) C14H17NO2

  • Molecular Weight 231.29

  •  MDL number MFCD05664133

  •  PubChem Substance ID 329778120

Purchase

Properties

Related Categories Building Blocks, C14, Chemical Synthesis, Ethers, Isocyanates,
SMILES string   O=C=N[C@@H]1CCCC[C@H]1OCc2ccccc2
InChI   1S/C14H17NO2/c16-11-15-13-8-4-5-9-14(13)17-10-12-6-2-1-3-7-12/h1-3,6-7,13-14H,4-5,8-10H2/t13-,14-/m1/s1
InChI key   HWVZRVVQFLEXHN-ZIAGYGMSSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Target organs 
Respiratory system
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?