• USA Home
  • CDS001296 - 5-Fluoro-N-methyl-2-phenoxybenzylamine

EMAIL THIS PAGE TO A FRIEND
CDS001296 Sigma-Aldrich

5-Fluoro-N-methyl-2-phenoxybenzylamine

AldrichCPR

  • Empirical Formula (Hill Notation) C14H14FNO

  • Molecular Weight 231.27

  •  MDL number MFCD08064241

  •  PubChem Substance ID 329778506

Purchase

Properties

Related Categories Aryl Fluorinated Building Blocks, Building Blocks, C13-C26, Chemical Synthesis, Fluorinated Building Blocks,
SMILES string   CNCc1cc(F)ccc1Oc2ccccc2
InChI   1S/C14H14FNO/c1-16-10-11-9-12(15)7-8-14(11)17-13-5-3-2-4-6-13/h2-9,16H,10H2,1H3
InChI key   JDHPXUQPTBDLHJ-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?