• USA Home
  • CDS001357 - 1-[3-(1-Piperidinylcarbonyl)-2-pyridinyl]piperazine

EMAIL THIS PAGE TO A FRIEND
CDS001357 Sigma-Aldrich

1-[3-(1-Piperidinylcarbonyl)-2-pyridinyl]piperazine

AldrichCPR

  • Empirical Formula (Hill Notation) C15H22N4O

  • Molecular Weight 274.36

  •  MDL number MFCD08061020

  •  PubChem Substance ID 329778553

Purchase

Properties

Related Categories Building Blocks, C13 to C15, C14 to C15, Chemical Synthesis, Heterocyclic Building Blocks,
SMILES string   O=C(N1CCCCC1)c2cccnc2N3CCNCC3
InChI   1S/C15H22N4O/c20-15(19-9-2-1-3-10-19)13-5-4-6-17-14(13)18-11-7-16-8-12-18/h4-6,16H,1-3,7-12H2
InChI key   WIVGCIGQDNMDQO-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?