• USA Home
  • CDS001365 - N-[1-(3-Chloro-2-pyridinyl)-4-piperidinyl]acetamide

EMAIL THIS PAGE TO A FRIEND
CDS001365 Sigma-Aldrich

N-[1-(3-Chloro-2-pyridinyl)-4-piperidinyl]acetamide

AldrichCPR

  • Empirical Formula (Hill Notation) C12H16ClN3O

  • Molecular Weight 253.73

  •  MDL number MFCD08060988

  •  PubChem Substance ID 329777154

Purchase

Properties

Related Categories Building Blocks, C12, C12 to C15, Chemical Synthesis, Halogenated Heterocycles,
SMILES string   CC(=O)NC1CCN(CC1)c2ncccc2Cl
InChI   1S/C12H16ClN3O/c1-9(17)15-10-4-7-16(8-5-10)12-11(13)3-2-6-14-12/h2-3,6,10H,4-5,7-8H2,1H3,(H,15,17)
InChI key   NOFVKQABVJSNPB-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?