• USA Home
  • CDS002397 - [1-(4-Ethoxybenzyl)-3-oxo-2-piperazinyl]acetic acid

EMAIL THIS PAGE TO A FRIEND
CDS002397 Sigma-Aldrich

[1-(4-Ethoxybenzyl)-3-oxo-2-piperazinyl]acetic acid

AldrichCPR

  • Empirical Formula (Hill Notation) C15H20N2O4

  • Molecular Weight 292.33

  •  MDL number MFCD05666718

  •  PubChem Substance ID 329779426

Purchase

Properties

Related Categories Building Blocks, C14 to C15, Chemical Synthesis, Heterocyclic Building Blocks, Piperazines More...
SMILES string   CCOc1ccc(CN2CCNC(=O)C2CC(O)=O)cc1
InChI   1S/C15H20N2O4/c1-2-21-12-5-3-11(4-6-12)10-17-8-7-16-15(20)13(17)9-14(18)19/h3-6,13H,2,7-10H2,1H3,(H,16,20)(H,18,19)
InChI key   UFAPTAYBYWFFKM-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
UN 2811 6.1 / PGIII

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?